Assay Details
| Gene Symbol: LOC100758550 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007640994.1 | XP_007639184.1 | 4 - 5 | 754 | 118 | |
| XM_007640995.1 | XP_007639185.1 | 4 - 5 | 868 | 118 | ||
| XM_007640996.1 | XP_007639186.1 | 3 - 4 | 736 | 118 | ||
More Information
Set Membership: |
|

LOC100758550
Genomic Map