Assay Details
| Gene Symbol: Isoc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007649781.1 | XP_007647971.1 | 2 - 3 | 511 | 59 | |
| GenBank mRNA | JI889038.1 | - | 2 - 3 | 262 | 59 | |
More Information
Set Membership: |
|

Isoc1
Genomic Map