Assay Details
| Gene Symbol: Fbxo44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003507041.2 | XP_003507089.1 | 1 - 2 | 264 | 69 | |
| XM_003507042.2 | XP_003507090.1 | 1 - 2 | 264 | 69 | ||
More Information
Set Membership: |
|

Fbxo44
Genomic Map