Assay Details
| Gene Symbol: Rnf5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003508463.2 | XP_003508511.1 | 5 - 6 | 512 | 91 | |
| GenBank mRNA | JI870543.1 | - | 5 - 6 | 431 | 91 | |
| JP051351.1 | - | 5 - 6 | 431 | 91 | ||
More Information
Set Membership: |
|

Rnf5
Genomic Map