Assay Details
| Gene Symbol: Ankrd23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007625771.1 | XP_007623961.1 | 7 - 8 | 758 | 63 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ankrd23| Gene Symbol: Ankrd23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007625771.1 | XP_007623961.1 | 7 - 8 | 758 | 63 | |
Set Membership: |
|