Assay Details
| Gene Symbol: Znf827 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007641492.1 | XP_007639682.1 | 8 - 9 | 2814 | 59 | |
| XM_007641493.1 | XP_007639683.1 | 8 - 9 | 2814 | 59 | ||
| XM_007641494.1 | XP_007639684.1 | 8 - 9 | 2346 | 59 | ||
| XM_007641495.1 | XP_007639685.1 | 8 - 9 | 2814 | 59 | ||
| XM_007641496.1 | XP_007639686.1 | 8 - 9 | 2814 | 59 | ||
| GenBank mRNA | JI881543.1 | - | 5 - 6 | 675 | 59 | |
| JP047126.1 | - | 5 - 6 | 675 | 59 | ||
More Information
Set Membership: |
|

Znf827
Genomic Map