Assay Details
| Gene Symbol: LOC100760824 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003494923.2 | XP_003494971.1 | 2 - 3 | 207 | 107 | |
| XM_007637791.1 | XP_007635981.1 | 2 - 3 | 207 | 107 | ||
| GenBank mRNA | JP046911.1 | - | 2 - 3 | 186 | 107 | |
More Information
Set Membership: |
|

LOC100760824
Genomic Map