Assay Details
| Gene Symbol: LOC100760824 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003494923.2 | XP_003494971.1 | 5 - 6 | 686 | 114 | |
| XM_007637791.1 | XP_007635981.1 | 5 - 6 | 686 | 114 | ||
| GenBank mRNA | JI875240.1 | - | 4 - 5 | 517 | 114 | |
| JP046911.1 | - | 5 - 6 | 665 | 114 | ||
More Information
Set Membership: |
|

LOC100760824
Genomic Map