Assay Details
| Gene Symbol: LOC100760824 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003494923.2 | XP_003494971.1 | 8 - 9 | 1005 | 95 | |
| GenBank mRNA | JI875240.1 | - | 7 - 8 | 836 | 95 | |
| JP046911.1 | - | 8 - 9 | 984 | 95 | ||
More Information
Set Membership: |
|

LOC100760824
Genomic Map