Assay Details
| Gene Symbol: Lipm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003505959.1 | XP_003506007.1 | 7 - 8 | 926 | 71 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lipm| Gene Symbol: Lipm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003505959.1 | XP_003506007.1 | 7 - 8 | 926 | 71 | |
Set Membership: |
|