Assay Details
| Gene Symbol: Atp5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003506176.2 | XP_003506224.1 | 4 - 5 | 646 | 100 | |
| GenBank mRNA | JI874525.1 | - | 3 - 4 | 411 | 100 | |
| JP060300.1 | - | 3 - 4 | 411 | 100 | ||
More Information
Set Membership: |
|

Atp5b
Genomic Map