Assay Details
| Gene Symbol: LOC100774629 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003510455.2 | XP_003510503.1 | 1 - 2 | 177 | 100 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100774629| Gene Symbol: LOC100774629 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003510455.2 | XP_003510503.1 | 1 - 2 | 177 | 100 | |
Set Membership: |
|