Assay Details
| Gene Symbol: Sdcbp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003501972.2 | XP_003502020.1 | 2 - 3 | 141 | 66 | |
| XM_007642594.1 | XP_007640784.1 | 2 - 3 | 141 | 66 | ||
| GenBank mRNA | JI892992.1 | - | 2 - 3 | 189 | 66 | |
| JP057451.1 | - | 2 - 3 | 189 | 66 | ||
More Information
Set Membership: |
|

Sdcbp
Genomic Map