Assay Details
| Gene Symbol: Tmem180 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007648683.1 | XP_007646873.1 | 6 - 7 | 1116 | 61 | |
| XM_007648684.1 | XP_007646874.1 | 6 - 7 | 1112 | 61 | ||
More Information
Set Membership: |
|

Tmem180
Genomic Map