Assay Details
| Gene Symbol: Lacc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003500302.2 | XP_003500350.1 | 3 - 4 | 947 | 71 | |
| XM_007641011.1 | XP_007639201.1 | 3 - 4 | 947 | 71 | ||
| XM_007641012.1 | XP_007639202.1 | 3 - 4 | 947 | 71 | ||
| XM_007641013.1 | XP_007639203.1 | 3 - 4 | 947 | 71 | ||
| GenBank mRNA | JI863040.1 | - | 3 - 4 | 904 | 71 | |
| JP057077.1 | - | 3 - 4 | 904 | 71 | ||
More Information
Set Membership: |
|

Lacc1
Genomic Map