Assay Details
| Gene Symbol: Hmox2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003506347.2 | XP_003506395.1 | 2 - 3 | 260 | 62 | |
| XM_007646859.1 | XP_007645049.1 | 3 - 4 | 334 | 62 | ||
| XM_007646860.1 | XP_007645050.1 | 2 - 3 | 220 | 62 | ||
| XM_007646861.1 | XP_007645051.1 | 2 - 3 | 199 | 62 | ||
| GenBank mRNA | JI895200.1 | - | 1 - 2 | 117 | 62 | |
| JP055370.1 | - | 3 - 4 | 295 | 62 | ||
More Information
Set Membership: |
|

Hmox2
Genomic Map