Assay Details
| Gene Symbol: Gpank1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003508483.2 | XP_003508531.1 | 2 - 3 | 804 | 67 | |
| XM_007649095.1 | XP_007647285.1 | 3 - 4 | 945 | 67 | ||
| GenBank mRNA | JI871406.1 | - | 1 - 2 | 263 | 67 | |
More Information
Set Membership: |
|

Gpank1
Genomic Map