Assay Details
| Gene Symbol: Prdx5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003511274.2 | XP_003511322.1 | 5 - 6 | 633 | 66 | |
| GenBank mRNA | JI872379.1 | - | 5 - 6 | 528 | 66 | |
| JP039713.1 | - | 5 - 6 | 476 | 66 | ||
More Information
Set Membership: |
|

Prdx5
Genomic Map