Assay Details
| Gene Symbol: Ube2q2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007619792.1 | XP_007617982.1 | 6 - 7 | 630 | 68 | |
| GenBank mRNA | JP039569.1 | - | 4 - 5 | 452 | 68 | |
More Information
Set Membership: |
|

Ube2q2
Genomic Map