Assay Details
| Gene Symbol: Trmt10a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007653858.1 | XP_007652048.1 | 4 - 5 | 592 | 58 | |
| GenBank mRNA | JI879135.1 | - | 4 - 5 | 619 | 58 | |
| JP064296.1 | - | 4 - 5 | 619 | 58 | ||
More Information
Set Membership: |
|

Trmt10a
Genomic Map