Assay Details
| Gene Symbol: Hn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007643039.1 | XP_007641229.1 | 1 - 2 | 165 | 99 | |
| GenBank mRNA | JI890390.1 | - | 1 - 2 | 310 | 99 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hn1| Gene Symbol: Hn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007643039.1 | XP_007641229.1 | 1 - 2 | 165 | 99 | |
| GenBank mRNA | JI890390.1 | - | 1 - 2 | 310 | 99 | |