Assay Details
| Gene Symbol: Ebp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003505107.2 | XP_003505155.1 | 2 - 3 | 473 | 71 | |
| GenBank mRNA | JI881952.1 | - | 1 - 2 | 366 | 71 | |
| JP065712.1 | - | 2 - 3 | 480 | 71 | ||
More Information
Set Membership: |
|

Ebp
Genomic Map