Assay Details
| Gene Symbol: Chi3l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003498875.2 | XP_003498923.2 | 9 - 10 | 1037 | 56 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Chi3l1| Gene Symbol: Chi3l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003498875.2 | XP_003498923.2 | 9 - 10 | 1037 | 56 | |
Set Membership: |
|