Assay Details
| Gene Symbol: March5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003498963.2 | XP_003499011.1 | 1 - 2 | 112 | 62 | |
| XM_007636098.1 | XP_007634288.1 | 1 - 2 | 112 | 62 | ||
More Information
Set Membership: |
|

March5
Genomic Map