Assay Details
| Gene Symbol: Mob1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003510050.2 | XP_003510098.1 | 3 - 4 | 512 | 91 | |
| XM_007650699.1 | XP_007648889.1 | 3 - 4 | 515 | 91 | ||
More Information
Set Membership: |
|

Mob1b
Genomic Map