Assay Details
| Gene Symbol: Gsx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003496674.1 | XP_003496722.1 | 4 - 5 | 854 | 73 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gsx1| Gene Symbol: Gsx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003496674.1 | XP_003496722.1 | 4 - 5 | 854 | 73 | |
Set Membership: |
|