Assay Details
| Gene Symbol: Eri1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003497955.2 | XP_003498003.1 | 4 - 5 | 490 | 67 | |
| GenBank mRNA | JI896563.1 | - | 3 - 4 | 347 | 67 | |
More Information
Set Membership: |
|

Eri1
Genomic Map