Assay Details
| Gene Symbol: Mob3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003502580.2 | XP_003502628.1 | 4 - 5 | 855 | 69 | |
| GenBank mRNA | JI861984.1 | - | 2 - 3 | 486 | 69 | |
| JP057276.1 | - | 2 - 3 | 486 | 69 | ||
More Information
Set Membership: |
|

Mob3a
Genomic Map