Assay Details
| Gene Symbol: Pdcl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003503328.2 | XP_003503376.1 | 5 - 6 | 658 | 66 | |
| GenBank mRNA | JI895590.1 | - | 5 - 6 | 670 | 66 | |
| JP045133.1 | - | 5 - 6 | 669 | 66 | ||
More Information
Set Membership: |
|

Pdcl3
Genomic Map