Assay Details
| Gene Symbol: Pus7l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003512134.2 | XP_003512182.2 | 7 - 8 | 1859 | 78 | |
| XM_007652620.1 | XP_007650810.1 | 7 - 8 | 1797 | 78 | ||
More Information
Set Membership: |
|

Pus7l
Genomic Map