Assay Details
| Gene Symbol: Ubtd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003497755.2 | XP_003497803.1 | 2 - 3 | 514 | 98 | |
| XM_007617220.1 | XP_007615410.1 | 2 - 3 | 277 | 98 | ||
| XM_007617227.1 | XP_007615417.1 | 2 - 3 | 327 | 98 | ||
| GenBank mRNA | JI866932.1 | - | 2 - 3 | 257 | 98 | |
More Information
Set Membership: |
|

Ubtd1
Genomic Map