Assay Details
| Gene Symbol: Foxn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003499538.2 | XP_003499586.1 | 3 - 4 | 809 | 89 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Foxn2| Gene Symbol: Foxn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003499538.2 | XP_003499586.1 | 3 - 4 | 809 | 89 | |
Set Membership: |
|