Assay Details
| Gene Symbol: Zdhhc7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003495111.2 | XP_003495159.1 | 2 - 3 | 414 | 63 | |
| XM_007655065.1 | XP_007653255.1 | 2 - 3 | 414 | 63 | ||
| XM_007655093.1 | XP_007653283.1 | 3 - 4 | 626 | 63 | ||
| XM_007655150.1 | XP_007653340.1 | 2 - 3 | 197 | 63 | ||
| GenBank mRNA | JI894215.1 | - | 2 - 3 | 405 | 63 | |
| JP059016.1 | - | 2 - 3 | 405 | 63 | ||
More Information
Set Membership: |
|

Zdhhc7
Genomic Map