Assay Details
| Gene Symbol: Mrpl28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003501498.2 | XP_003501546.2 | 3 - 4 | 691 | 74 | |
| XM_007642127.1 | XP_007640317.1 | 2 - 3 | 538 | 74 | ||
| GenBank mRNA | JI879461.1 | - | 3 - 4 | 624 | 74 | |
| JP042728.1 | - | 3 - 4 | 618 | 74 | ||
More Information
Set Membership: |
|

Mrpl28
Genomic Map