Assay Details
| Gene Symbol: Smim14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003512235.2 | XP_003512283.1 | 3 - 4 | 226 | 70 | |
| XM_007652737.1 | XP_007650927.1 | 3 - 4 | 318 | 70 | ||
| GenBank mRNA | JI861942.1 | - | 3 - 4 | 281 | 70 | |
| JP061008.1 | - | 3 - 4 | 281 | 70 | ||
More Information
Set Membership: |
|

Smim14
Genomic Map