Assay Details
| Gene Symbol: Pebp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007647830.1 | XP_007646020.1 | 1 - 2 | 83 | 60 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pebp4| Gene Symbol: Pebp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007647830.1 | XP_007646020.1 | 1 - 2 | 83 | 60 | |
Set Membership: |
|