Assay Details
| Gene Symbol: Arf5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007613346.1 | XP_007611536.1 | 1 - 2 | 124 | 121 | |
| GenBank mRNA | JP054225.1 | - | 1 - 2 | 124 | 121 | |
More Information
Set Membership: |
|

Arf5
Genomic Map