Assay Details
| Gene Symbol: Ndufb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007643652.1 | XP_007641842.1 | 2 - 3 | 339 | 66 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ndufb2| Gene Symbol: Ndufb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007643652.1 | XP_007641842.1 | 2 - 3 | 339 | 66 | |
Set Membership: |
|