Assay Details
| Gene Symbol: Mrps30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007627282.1 | XP_007625472.1 | 4 - 5 | 730 | 79 | |
| GenBank mRNA | JI892086.1 | - | 3 - 4 | 406 | 79 | |
| JP053535.1 | - | 3 - 4 | 406 | 79 | ||
More Information
Set Membership: |
|

Mrps30
Genomic Map