Assay Details
| Gene Symbol: Lyplal1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003494999.2 | XP_003495047.1 | 1 - 2 | 111 | 78 | |
| GenBank mRNA | JP065359.1 | - | 1 - 2 | 213 | 78 | |
More Information
Set Membership: |
|

Lyplal1
Genomic Map