Assay Details
| Gene Symbol: LOC100758957 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003506639.2 | XP_003506687.1 | 90 - 91 | 14073 | 63 | |
| GenBank mRNA | JI887932.1 | - | 4 - 5 | 527 | 63 | |
More Information
Set Membership: |
|

LOC100758957
Genomic Map