Assay Details
| Gene Symbol: Gnrh1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003496189.2 | XP_003496237.2 | 1 - 2 | 1673 | 64 | |
| XM_007647787.1 | XP_007645977.1 | 1 - 2 | 1673 | 64 | ||
More Information
Set Membership: |
|

Gnrh1
Genomic Map