Assay Details
| Gene Symbol: Mrpl9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007653333.1 | XP_007651523.1 | 2 - 3 | 342 | 74 | |
| GenBank mRNA | JI868407.1 | - | 2 - 3 | 318 | 74 | |
More Information
Set Membership: |
|

Mrpl9
Genomic Map