Assay Details
| Gene Symbol: Vstm4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003495349.2 | XP_003495397.1 | 7 - 8 | 847 | 66 | |
| GenBank mRNA | JI896841.1 | - | 5 - 6 | 382 | 66 | |
More Information
Set Membership: |
|

Vstm4
Genomic Map