Assay Details
| Gene Symbol: Znf704 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003513387.1 | XP_003513435.1 | 5 - 6 | 804 | 60 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Znf704| Gene Symbol: Znf704 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003513387.1 | XP_003513435.1 | 5 - 6 | 804 | 60 | |
Set Membership: |
|