Assay Details
| Gene Symbol: Fbxo5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007644909.1 | XP_007643099.1 | 3 - 4 | 3475 | 65 | |
| XM_007644911.1 | XP_007643101.1 | 4 - 5 | 1143 | 65 | ||
| GenBank mRNA | JP061112.1 | - | 3 - 4 | 968 | 65 | |
More Information
Set Membership: |
|

Fbxo5
Genomic Map