Assay Details
| Gene Symbol: Pin4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007649429.1 | XP_007647619.1 | 1 | 223 | 82 | |
| GenBank mRNA | JI873223.1 | - | 1 | 223 | 82 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pin4| Gene Symbol: Pin4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007649429.1 | XP_007647619.1 | 1 | 223 | 82 | |
| GenBank mRNA | JI873223.1 | - | 1 | 223 | 82 | |
Set Membership: |
|