Assay Details
| Gene Symbol: Kctd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007643375.1 | XP_007641565.1 | 5 - 6 | 813 | 80 | |
| GenBank mRNA | JI895503.1 | - | 1 - 2 | 76 | 80 | |
More Information
Set Membership: |
|

Kctd2
Genomic Map