Assay Details
| Gene Symbol: Narfl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003501562.2 | XP_003501610.1 | 5 - 6 | 617 | 60 | |
| GenBank mRNA | JP043347.1 | - | 1 - 2 | 42 | 60 | |
More Information
Set Membership: |
|

Narfl
Genomic Map