Assay Details
| Gene Symbol: Med4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003508406.2 | XP_003508454.1 | 7 - 8 | 971 | 81 | |
| XM_007649038.1 | XP_007647228.1 | 6 - 7 | 936 | 81 | ||
| GenBank mRNA | JI886589.1 | - | 7 - 8 | 973 | 81 | |
More Information
Set Membership: |
|

Med4
Genomic Map